Skip to Content
Merck
All Photos(1)

Key Documents

ENA580356254

Enamine

{1-oxaspiro[4.5]decan-2-yl}methanol

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CZK 8,730.00
100 MG
CZK 12,467.50
250 MG
CZK 17,310.00
500 MG
CZK 26,605.00
1 G
CZK 33,820.00
2.5 G
CZK 65,215.00
5 G
CZK 95,960.00

Vendor SKU: EN300-328895-50MG

CZK 8,730.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
CZK 8,730.00
100 MG
CZK 12,467.50
250 MG
CZK 17,310.00
500 MG
CZK 26,605.00
1 G
CZK 33,820.00
2.5 G
CZK 65,215.00
5 G
CZK 95,960.00

About This Item

CAS Number:
Molecular Weight:
170.25
MDL number:

Vendor SKU: EN300-328895-50MG

CZK 8,730.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

(1-oxaspiro[4.5]decan-2-yl)methanol

Inchi Code

1S/C10H18O2/c11-8-9-4-7-10(12-9)5-2-1-3-6-10/h9,11H,1-8H2

InChI key

NXROIUACPSTQQX-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-328895

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-328895


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service