Skip to Content
Merck
All Photos(1)

Key Documents

ENA458425001

Enamine

(3-amino-1,1,1-trifluoropropan-2-yl)(benzyl)amine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 10,907.50
250 MG
CZK 15,100.00
500 MG
CZK 23,192.50
1 G
CZK 29,400.00
2.5 G
CZK 56,537.50
5 G
CZK 83,122.50

Vendor SKU: EN300-100043-100MG

CZK 10,907.50


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CZK 10,907.50
250 MG
CZK 15,100.00
500 MG
CZK 23,192.50
1 G
CZK 29,400.00
2.5 G
CZK 56,537.50
5 G
CZK 83,122.50

About This Item

CAS Number:
Molecular Weight:
218.22
MDL number:

Vendor SKU: EN300-100043-100MG

CZK 10,907.50


Ships in 14 days - from Aldrich Partner.

Iupac Name

N2-benzyl-3,3,3-trifluoropropane-1,2-diamine

Inchi Code

1S/C10H13F3N2/c11-10(12,13)9(6-14)15-7-8-4-2-1-3-5-8/h1-5,9,15H,6-7,14H2

InChI key

ZVQBRWKRJPPJQF-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-100043

MSDS

https://www.enaminestore.com/catalog/EN300-100043


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service