Skip to Content
Merck
All Photos(1)

Key Documents

ENA423209553

Enamine

6-(pyrazin-2-ylsulfanyl)pyridine-2-carbonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 10,355.00
250 MG
CZK 14,287.50
500 MG
CZK 21,795.00
1 G
CZK 27,645.00
2.5 G
CZK 53,092.50
5 G
CZK 78,020.00

Vendor SKU: EN300-170387-100MG

CZK 10,355.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CZK 10,355.00
250 MG
CZK 14,287.50
500 MG
CZK 21,795.00
1 G
CZK 27,645.00
2.5 G
CZK 53,092.50
5 G
CZK 78,020.00

About This Item

CAS Number:
Molecular Weight:
214.25
MDL number:

Vendor SKU: EN300-170387-100MG

CZK 10,355.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

6-(2-pyrazinylsulfanyl)-2-pyridinecarbonitrile

Inchi Code

1S/C10H6N4S/c11-6-8-2-1-3-9(14-8)15-10-7-12-4-5-13-10/h1-5,7H

InChI key

QERSAHRCGCWUKU-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-170387

MSDS

https://www.enaminestore.com/catalog/EN300-170387


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service