Skip to Content
Merck
All Photos(1)

Key Documents

ENA405194271

Enamine

1-phenylpropane-1,3-diamine

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 17,595.00
250 MG
CZK 22,080.00
500 MG
CZK 30,757.50
1 G
CZK 37,485.00

Vendor SKU: EN300-133488-100MG

CZK 17,595.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CZK 17,595.00
250 MG
CZK 22,080.00
500 MG
CZK 30,757.50
1 G
CZK 37,485.00

About This Item

CAS Number:
Molecular Weight:
150.22
MDL number:

Vendor SKU: EN300-133488-100MG

CZK 17,595.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

1-phenyl-1,3-propanediamine

Inchi Code

1S/C9H14N2/c10-7-6-9(11)8-4-2-1-3-5-8/h1-5,9H,6-7,10-11H2

InChI key

PLDPCRJUQLPMNU-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-133488

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-133488


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service