Skip to Content
Merck
All Photos(1)

Key Documents

ENA387772867

Enamine

7,9-dimethyl-1,3-diazaspiro[4.5]decane-2,4-dione

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 9,412.50
250 MG
CZK 12,955.00
500 MG
CZK 19,715.00
1 G
CZK 24,980.00
2.5 G
CZK 47,860.00
5 G
CZK 70,285.00
10 G
CZK 103,727.50

Vendor SKU: EN300-109179-100MG

CZK 9,412.50


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CZK 9,412.50
250 MG
CZK 12,955.00
500 MG
CZK 19,715.00
1 G
CZK 24,980.00
2.5 G
CZK 47,860.00
5 G
CZK 70,285.00
10 G
CZK 103,727.50

About This Item

CAS Number:
Molecular Weight:
196.25
MDL number:

Vendor SKU: EN300-109179-100MG

CZK 9,412.50


Ships in 14 days - from Aldrich Partner.

mp

333-334

Iupac Name

7,9-dimethyl-1,3-diazaspiro[4.5]decane-2,4-dione

Inchi Code

1S/C10H16N2O2/c1-6-3-7(2)5-10(4-6)8(13)11-9(14)12-10/h6-7H,3-5H2,1-2H3,(H2,11,12,13,14)

InChI key

YYLYCNHLXYNSSJ-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-109179

Melting Point

333-334

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-109179


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service