Skip to Content
Merck
All Photos(1)

Key Documents

ENA370563267

Enamine

methyl 2-amino-3-methoxy-2-methylpropanoate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 4,992.50
250 MG
CZK 6,617.50
500 MG
CZK 11,395.00
1 G
CZK 14,807.50
2.5 G
CZK 27,937.50
5 G
CZK 40,775.00
10 G
CZK 59,917.50

Vendor SKU: EN300-105490-100MG

CZK 4,992.50


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CZK 4,992.50
250 MG
CZK 6,617.50
500 MG
CZK 11,395.00
1 G
CZK 14,807.50
2.5 G
CZK 27,937.50
5 G
CZK 40,775.00
10 G
CZK 59,917.50

About This Item

CAS Number:
Molecular Weight:
147.17
MDL number:

Vendor SKU: EN300-105490-100MG

CZK 4,992.50


Ships in 14 days - from Aldrich Partner.

Iupac Name

methyl 2-amino-3-methoxy-2-methylpropanoate

Inchi Code

1S/C6H13NO3/c1-6(7,4-9-2)5(8)10-3/h4,7H2,1-3H3

InChI key

DCTDZOZFFDUTRP-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-105490

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-105490


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service