Skip to Content
Merck
All Photos(1)

Key Documents

ENA333557795

Enamine

2-[(3-bromopropoxy)methyl]oxolane

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 8,925.00
250 MG
CZK 12,272.50

Vendor SKU: EN300-81928-100MG

CZK 8,925.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CZK 8,925.00
250 MG
CZK 12,272.50

About This Item

CAS Number:
Molecular Weight:
223.11
MDL number:

Vendor SKU: EN300-81928-100MG

CZK 8,925.00


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-[(3-bromopropoxy)methyl]tetrahydrofuran

Inchi Code

1S/C8H15BrO2/c9-4-2-5-10-7-8-3-1-6-11-8/h8H,1-7H2

InChI key

BPBXDWYCBLYUJH-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-81928

MSDS

https://www.enaminestore.com/catalog/EN300-81928


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service