Skip to Content
Merck
All Photos(1)

Key Documents

ENA249152257

Enamine

2-(carbamoylamino)hexanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 3,660.00
250 MG
CZK 4,700.00
500 MG
CZK 7,885.00
1 G
CZK 10,810.00
2.5 G
CZK 20,105.00
5 G
CZK 29,237.50
10 G
CZK 42,822.50

Vendor SKU: EN300-55744-100MG

CZK 3,660.00


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CZK 3,660.00
250 MG
CZK 4,700.00
500 MG
CZK 7,885.00
1 G
CZK 10,810.00
2.5 G
CZK 20,105.00
5 G
CZK 29,237.50
10 G
CZK 42,822.50

About This Item

CAS Number:
Molecular Weight:
174.2
MDL number:

Vendor SKU: EN300-55744-100MG

CZK 3,660.00


Ships in 14 days - from Aldrich Partner.

mp

169-170

Iupac Name

N-(aminocarbonyl)norleucine

Inchi Code

1S/C7H14N2O3/c1-2-3-4-5(6(10)11)9-7(8)12/h5H,2-4H2,1H3,(H,10,11)(H3,8,9,12)

InChI key

CRTBNFJSWPZSQM-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-55744

Melting Point

169-170

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-55744


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service