Skip to Content
Merck
All Photos(1)

Key Documents

ENA112933395

Enamine

2-(carbamoylamino)-2-phenylacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 5,187.50
250 MG
CZK 6,942.50
500 MG
CZK 12,077.50
1 G
CZK 15,685.00
2.5 G
CZK 29,692.50
5 G
CZK 43,342.50
10 G
CZK 63,752.50

Vendor SKU: EN300-26540-100MG

CZK 5,187.50


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
100 MG
CZK 5,187.50
250 MG
CZK 6,942.50
500 MG
CZK 12,077.50
1 G
CZK 15,685.00
2.5 G
CZK 29,692.50
5 G
CZK 43,342.50
10 G
CZK 63,752.50

About This Item

CAS Number:
Molecular Weight:
194.19
MDL number:

Vendor SKU: EN300-26540-100MG

CZK 5,187.50


Ships in 14 days - from Aldrich Partner.

mp

200-205

Iupac Name

[(aminocarbonyl)amino](phenyl)acetic acid

Inchi Code

1S/C9H10N2O3/c10-9(14)11-7(8(12)13)6-4-2-1-3-5-6/h1-5,7H,(H,12,13)(H3,10,11,14)

InChI key

GIOUOHDKHHZWIQ-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-26540

Melting Point

200-205

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-26540


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service