Skip to Content
Merck
All Photos(1)

Key Documents

ENA024295099

Enamine

4-[(1,1-dioxo-1lambda6,2-benzothiazol-3-yl)amino]butanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 1,690.00
250 MG
CZK 2,437.50
500 MG
CZK 3,835.00
1 G
CZK 4,907.50
2.5 G
CZK 9,717.50
5 G
CZK 14,397.50
10 G
CZK 21,287.50

Vendor SKU: EN300-09061-100MG

CZK 1,690.00


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 MG
CZK 1,690.00
250 MG
CZK 2,437.50
500 MG
CZK 3,835.00
1 G
CZK 4,907.50
2.5 G
CZK 9,717.50
5 G
CZK 14,397.50
10 G
CZK 21,287.50

About This Item

CAS Number:
Molecular Weight:
268.29
MDL number:

Vendor SKU: EN300-09061-100MG

CZK 1,690.00


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

mp

200-202

Iupac Name

4-[(1,1-dioxido-1,2-benzisothiazol-3-yl)amino]butanoic acid

Inchi Code

1S/C11H12N2O4S/c14-10(15)6-3-7-12-11-8-4-1-2-5-9(8)18(16,17)13-11/h1-2,4-5H,3,6-7H2,(H,12,13)(H,14,15)

InChI key

OKRQABOQRFCMRS-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-09061

Melting Point

200-202

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-09061


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service