Skip to Content
Merck
All Photos(1)

Key Documents

ENA018109127

Enamine

5-methyl-2-(trifluoromethyl)-4H,7H-[1,2,4]triazolo[1,5-a]pyrimidin-7-one

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 3,867.50
250 MG
CZK 5,525.00
500 MG
CZK 8,710.00
1 G
CZK 11,180.00
2.5 G
CZK 23,497.50
5 G
CZK 43,972.50
10 G
CZK 62,562.50

Vendor SKU: EN300-05910-100MG

CZK 3,867.50


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 MG
CZK 3,867.50
250 MG
CZK 5,525.00
500 MG
CZK 8,710.00
1 G
CZK 11,180.00
2.5 G
CZK 23,497.50
5 G
CZK 43,972.50
10 G
CZK 62,562.50

About This Item

CAS Number:
Molecular Weight:
218.14
MDL number:

Vendor SKU: EN300-05910-100MG

CZK 3,867.50


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

mp

265-267

Iupac Name

5-methyl-2-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidin-7-ol

Inchi Code

1S/C7H5F3N4O/c1-3-2-4(15)14-6(11-3)12-5(13-14)7(8,9)10/h2H,1H3,(H,11,12,13)

InChI key

RCFXSBDEUTWHMQ-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-05910

Melting Point

265-267

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-05910


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service