COM448667319
Combi-Blocks, Inc.
1-Anthracenecarboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1-anthracenecarboxylic acid
Inchi Code
1S/C15H10O2/c16-15(17)13-7-3-6-12-8-10-4-1-2-5-11(10)9-14(12)13/h1-9H,(H,16,17)
InChI key
CCFAKBRKTKVJPO-UHFFFAOYSA-N
Purity
98%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?QF-9963
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?QF-9963
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service