Skip to Content
Merck
All Photos(1)

Key Documents

CIAH987EB8B6

ChemScene LLC (Preferred Partner)

3-Bromo-1H-indole-6-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
240.06
MDL number:
Pricing and availability is not currently available. Contact Technical Service

bp

470.9C at 760 mmHg

Iupac Name

3-bromo-1H-indole-6-carboxylic acid

Inchi Code

1S/C9H6BrNO2/c10-7-4-11-8-3-5(9(12)13)1-2-6(7)8/h1-4,11H,(H,12,13)

InChI key

XZNVEIYDUGPMCS-UHFFFAOYSA-N

storage temp.

-20C, sealed storage, away from moisture

Purity

96%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/219508-19-7.html

Boiling point

470.9C at 760 mmHg

Pictograms

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0044446.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service