Skip to Content
Merck
All Photos(1)

Key Documents

BL3H1F1CA402

BLD Pharmatech Co., Ltd.

Pyrazin-2-ylmethanamine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CZK 1,718.75
1 G
CZK 1,891.25
5 G
CZK 4,737.50
10 G
CZK 8,043.75
25 G
CZK 18,480.00

Vendor SKU: BD46810-250MG

CZK 1,718.75


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
CZK 1,718.75
1 G
CZK 1,891.25
5 G
CZK 4,737.50
10 G
CZK 8,043.75
25 G
CZK 18,480.00

About This Item

CAS Number:
Molecular Weight:
145.59
MDL number:

Vendor SKU: BD46810-250MG

CZK 1,718.75


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-pyrazinylmethanamine hydrochloride

Inchi Code

1S/C5H7N3.ClH/c6-3-5-4-7-1-2-8-5;/h1-2,4H,3,6H2;1H

InChI key

FVGCPPSBZHDZPC-UHFFFAOYSA-N

storage temp.

Inert atmosphere,Room Temperature

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow to Brown Solid

Product Link

http://www.bldpharm.com/products/39204-49-4.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://file.bldpharm.com/static/upload/prosds/bldsds/47/SDS-BD46810.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service