Skip to Content
Merck
All Photos(1)

Key Documents

ATEH97F46ECD

AstaTech, Inc.

THIETAN-3-ONE 1,1-DIOXIDE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
120.13
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

3-thietanone 1,1-dioxide

Inchi Code

1S/C3H4O3S/c4-3-1-7(5,6)2-3/h1-2H2

InChI key

AYVTTWZAVAGWEX-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=187827

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=D75198


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service