Skip to Content
Merck
All Photos(1)

Key Documents

ATE002883202

AstaTech, Inc.

1-N-ALLOC-4-PIPERIDONE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
183.21
MDL number:

Iupac Name

allyl 4-oxo-1-piperidinecarboxylate

Inchi Code

1S/C9H13NO3/c1-2-7-13-9(12)10-5-3-8(11)4-6-10/h2H,1,3-7H2

InChI key

TUEIRODWNWORRI-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

96%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Colorless to Yellow Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=9793

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=A56036


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service