APO725205573
Apollo Scientific Ltd
Methyl 2-fluoro-4-(methylsulphonyl)benzoate
About This Item
Iupac Name
methyl 2-fluoro-4-(methylsulfonyl)benzoate
Inchi Code
1S/C9H9FO4S/c1-14-9(11)7-4-3-6(5-8(7)10)15(2,12)13/h3-5H,1-2H3
InChI key
NLLKCUOCUBCOFS-UHFFFAOYSA-N
storage temp.
Country of Origin
Shipping Temperature
Physical Form
Product Link
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/methyl-2-fluoro-4-methylsulphonylbenzoate
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service