Skip to Content
Merck
All Photos(1)

Key Documents

APO455827333

Apollo Scientific Ltd

N-(3-Formylphenyl)methanesulphonamide

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

CAS Number:
Molecular Weight:
199.23
MDL number:

bp

146-147

Iupac Name

N-(3-formylphenyl)methanesulfonamide

Inchi Code

1S/C8H9NO3S/c1-13(11,12)9-8-4-2-3-7(5-8)6-10/h2-6,9H,1H3

InChI key

CBDSSTWZEANOCR-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/n-3-formylphenylmethanesulphonamide

Boiling point

146-147

Pictograms

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/n-3-formylphenylmethanesulphonamide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service