CF0045
Ethyl difluoroazidoacetate
Synonym(s):
2-azido-2,2-difluoroacetic acid ethyl ester, Ethyl 2-azido-2,2-difluoroacetate
About This Item
Recommended Products
Quality Level
SMILES string
O=C(OCC)C(N=[N+]=[N-])(F)F
InChI
1S/C4H5F2N3O2/c1-2-11-3(10)4(5,6)8-9-7/h2H2,1H3
InChI key
BISKRTIGSBFIQH-UHFFFAOYSA-N
Related Categories
General description
Learn more about fluorinated click chemistry
Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Fluorinated azides are a new tool that can be used to introduce fluorinated groups to molecules. These molecules can utilize click chemistry to easily add fluorinated groups.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service