Skip to Content
Merck
All Photos(1)

Documents

901584

Sigma-Aldrich

15-Crown-5 solution

1 M in THF

Synonym(s):

1,4,7,10,13-Pentaoxacyclopentadecane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H20O5
CAS Number:
MDL number:
UNSPSC Code:
12352200

form

liquid

concentration

1 M in THF

density

0.930 g/mL

storage temp.

room temp

InChI

1S/C10H20O5/c1-2-12-5-6-14-9-10-15-8-7-13-4-3-11-1/h1-10H2

InChI key

VFTFKUDGYRBSAL-UHFFFAOYSA-N

Application

Valuable reagent provided in tetrahydrofuran for more convenient handling.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.0 °F

Flash Point(C)

-18.88 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service