Skip to Content
Merck
All Photos(1)

Key Documents

542830

Sigma-Aldrich

3,5-Bis(trifluoromethyl)benzamidoxime

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2C6H3C(NH2)=NOH
CAS Number:
Molecular Weight:
272.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

129-132 °C (lit.)

SMILES string

N\C(=N\O)c1cc(cc(c1)C(F)(F)F)C(F)(F)F

InChI

1S/C9H6F6N2O/c10-8(11,12)5-1-4(7(16)17-18)2-6(3-5)9(13,14)15/h1-3,18H,(H2,16,17)

InChI key

GGAPMNUHESBFNK-UHFFFAOYSA-N

General description

3,5-Bis(trifluoromethyl)benzamidoxime [N′-hydroxy-3,5-bis(trifluoromethyl)benzenecarboximidamide] is a BAO [3,5-bis(trifluoromethyl)] derivative containing amidoxime as a functional group].

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel and specific fluorescence reaction for uracil
Shibata, Takayuki, et al.
Analytica Chimica Acta, 674.2, 234- 238 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service