Skip to Content
Merck
All Photos(1)

Documents

539759

Sigma-Aldrich

8-Benzyloxyisosorbide triphenylphosphoylide solution

0.05 M in THF

Synonym(s):

Carb[(1R,4S,5R,8R)-8-benzyloxy-2,6-dioxabicyclo[3.3.0]octan-4-atemethylene]triphenylphosphorane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H31O5P
Molecular Weight:
538.57
UNSPSC Code:
12352005
PubChem Substance ID:

optical activity

[α]20/D +1.7°, neat

concentration

0.05 M in THF

refractive index

n20/D 1.413

bp

70-71 °C

density

0.882 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

O=C(O[C@H]1CO[C@@H]2[C@@H](CO[C@H]12)OCc3ccccc3)C=P(c4ccccc4)(c5ccccc5)c6ccccc6

InChI

1S/C33H31O5P/c34-31(38-30-23-37-32-29(22-36-33(30)32)35-21-25-13-5-1-6-14-25)24-39(26-15-7-2-8-16-26,27-17-9-3-10-18-27)28-19-11-4-12-20-28/h1-20,24,29-30,32-33H,21-23H2/t29-,30+,32-,33-/m1/s1

InChI key

RFSVJVARGBBXNB-NKKCXFSRSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F

Flash Point(C)

-17.0 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service