Skip to Content
Merck
All Photos(1)

Key Documents

42170

Sigma-Aldrich

2,4-Dinitrophenol

technical, moistened with water, ≥97.0% (HPLC)

Synonym(s):

α-Dinitrophenol, 2,4-DNP, DNP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3OH
CAS Number:
Molecular Weight:
184.11
Beilstein:
1246142
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

6.35 (vs air)

grade

technical

Assay

≥97.0% (HPLC)

quality

moistened with water

contains

~33% water as stabilizer

mp

108-112 °C (lit.)
108-112 °C

SMILES string

Oc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H4N2O5/c9-6-2-1-4(7(10)11)3-5(6)8(12)13/h1-3,9H

InChI key

UFBJCMHMOXMLKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Acute 1 - Desen. Expl. 1 - STOT RE 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammed Kaplan et al.
Nature methods, 12(7), 649-652 (2015-05-20)
Studying biomolecules at atomic resolution in their native environment is the ultimate aim of structural biology. We investigated the bacterial type IV secretion system core complex (T4SScc) by cellular dynamic nuclear polarization-based solid-state nuclear magnetic resonance spectroscopy to validate a
Bonnie L Westra et al.
Journal of the American Medical Informatics Association : JAMIA, 22(3), 600-607 (2015-02-12)
There is wide recognition that, with the rapid implementation of electronic health records (EHRs), large data sets are available for research. However, essential standardized nursing data are seldom integrated into EHRs and clinical data repositories. There are many diverse activities
Stephanie C Weber et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(19), 7338-7343 (2012-04-21)
Chromosomal loci jiggle in place between segregation events in prokaryotic cells and during interphase in eukaryotic nuclei. This motion seems random and is often attributed to brownian motion. However, we show here that locus dynamics in live bacteria and yeast
Toni-Maree Rogers et al.
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer, 10(4), 611-618 (2015-03-20)
The use of targeted therapies toward specific oncogenic driver mutations has become a critical factor in the treatment of patients with lung cancer. It is therefore essential to utilize tests with high performance characteristics. Fluorescence in situ hybridization (FISH) is
Sean Marrache et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(40), 16288-16293 (2012-09-20)
Mitochondrial dysfunctions cause numerous human disorders. A platform technology based on biodegradable polymers for carrying bioactive molecules to the mitochondrial matrix could be of enormous potential benefit in treating mitochondrial diseases. Here we report a rationally designed mitochondria-targeted polymeric nanoparticle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service