Skip to Content
Merck
All Photos(1)

Key Documents

405264

Sigma-Aldrich

Hexafluoro-2-butyne

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C≡CCF3
CAS Number:
Molecular Weight:
162.03
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

vapor pressure

105 psi ( 20 °C)

Assay

99%

bp

−25 °C (lit.)

mp

−117 °C (lit.)

SMILES string

FC(F)(F)C#CC(F)(F)F

InChI

1S/C4F6/c5-3(6,7)1-2-4(8,9)10

InChI key

WBCLXFIDEDJGCC-UHFFFAOYSA-N

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Pictograms

Gas cylinderSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service