Skip to Content
Merck
All Photos(1)

Key Documents

37342

Sigma-Aldrich

(1R,2S)-cis-1,2-Dihydro-1,2-naphthalenediol

≥98.0%

Synonym(s):

(1R-cis)-1,2-Dihydro-1,2-naphthalenediol, (1R,2S)-cis-1,2-Dihydro-1,2-dihydroxynaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
Beilstein:
1909468
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Assay

≥98.0%

optical activity

[α]20/D +242±5°, c = 1% in chloroform stab. with amylenes

mp

121-127 °C (lit.)
136-139 °C

storage temp.

2-8°C

SMILES string

O[C@H]1C=Cc2ccccc2[C@H]1O

InChI

1S/C10H10O2/c11-9-6-5-7-3-1-2-4-8(7)10(9)12/h1-6,9-12H/t9-,10+/m0/s1

InChI key

QPUHWUSUBHNZCG-VHSXEESVSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Chiral building block for preparing both enantiomers of 1,2-epoxy-1,2,3,4-tetrahydro- naphthalene

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Débora Maria Abrantes Costa et al.
Protein expression and purification, 132, 50-59 (2017-01-17)
The second enzyme of the naphthalene degradation pathway in Pseudomonas putida G7 is NahB, a dehydrogenase that converts cis-1,2-dihydroxy-1,2-dihydronaphthalene to 1,2-dihydroxynaphthalene. We report the cloning, optimization of expression, purification, kinetic studies and preliminary structural characterization of the recombinant NahB. The
A.J. Blacker et al.
PCT Int. Appl. 94, 19338 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service