Skip to Content
Merck
All Photos(1)

Key Documents

291021

Sigma-Aldrich

Iodine monobromide solution

1.0 M in methylene chloride

Synonym(s):

Bromine monoiodide, Bromoiodide, Iodine bromide, Iodobromine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IBr
CAS Number:
Molecular Weight:
206.81
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

concentration

1.0 M in methylene chloride

density

1.458 g/mL at 25 °C

SMILES string

BrI

InChI

1S/BrI/c1-2

InChI key

CBEQRNSPHCCXSH-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

L E Mikhno et al.
Likars'ka sprava, (1)(1), 89-91 (1992-01-01)
The effect of local iodine-bromine baths at the early stage of health-resort treatment on the main values of the cardio-vascular system was studied in patients suffering of myocardial infarction with arterial hypertension. It was established that local iodine-bromine baths produce
Ultraviolet spectrophotometric determination of phenols in natural and waste waters with iodine monobromide.
F Bosch et al.
The Analyst, 112(9), 1335-1337 (1987-09-01)
L P Markina et al.
Voprosy kurortologii, fizioterapii, i lechebnoi fizicheskoi kultury, (4)(4), 38-40 (1998-12-18)
The studies of 116 females with chronic nonspecific salpingo-oophoritis demonstrate that combination of low-frequency magnetic field with iodine-bromine water effectively corrects immunological indices, i.e. improves T- and B-cell immunity, production of IgM and IgA, proportion of immunoregulatory subpopulations of T-lymphocytes.
Z R Zunnunov
Terapevticheskii arkhiv, 82(1), 24-27 (2010-04-07)
to evaluate the efficiency of iodide-bromine balneotherapy (IBB) in patients with coronary heart disease (CHD) concurrent with chronic obstructive pulmonary disease (COPD). Sixty-six patients with CHD, Functional Class HII stable angina on exertion concurrent with mild and moderate COPD were
[A method for preparing iodine and iodine bromide baths with the presence of molecular iodine].
I I Gusarov et al.
Voprosy kurortologii, fizioterapii, i lechebnoi fizicheskoi kultury, (2)(2), 62-63 (1989-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service