Skip to Content
Merck
All Photos(1)

Documents

196908

Sigma-Aldrich

Perfluoroglutaric acid

97%

Synonym(s):

Hexafluoroglutaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CF2)3COOH
CAS Number:
Molecular Weight:
240.06
Beilstein:
1801049
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

97%

bp

134-138 °C/3 mmHg (lit.)

mp

88-91 °C (lit.)

SMILES string

OC(=O)C(F)(F)C(F)(F)C(F)(F)C(O)=O

InChI

1S/C5H2F6O4/c6-3(7,1(12)13)5(10,11)4(8,9)2(14)15/h(H,12,13)(H,14,15)

InChI key

CCUWGJDGLACFQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service