Přejít k obsahu
Merck
  • Parameterization of phosphine ligands demonstrates enhancement of nickel catalysis via remote steric effects.
Parameterization of phosphine ligands demonstrates enhancement of nickel catalysis via remote steric effects.
Nature chemistry (2017-07-30)
Kevin Wu, Abigail G Doyle
ANOTACE
The field of Ni-catalysed cross-coupling has seen rapid recent growth because of the low cost of Ni, its earth abundance, and its ability to promote unique cross-coupling reactions. Whereas advances in the related field of Pd-catalysed cross-coupling have been driven by ligand design, the development of ligands specifically for Ni has received minimal attention. Here, we disclose a class of phosphines that enable the Ni-catalysed Csp
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
TyrannoPhos