Přejít k obsahu
Merck
In vitro flow cytometry method to quantitatively assess inhibitors of P-glycoprotein.
Drug metabolism and disposition: the biological fate of chemicals (2000-04-20)
E J Wang, C N Casciano, R P Clement, W W Johnson
ANOTACE
P-glycoprotein (Pgp)-mediated drug efflux is a major factor contributing to the variance of absorption and distribution of many drugs. A simple and reliable in vitro method to identify inhibitors of Pgp helps to prevent the potential of drug interactions. Using daunorubicin as a fluorescent marker and vanadate as a positive control compound, a functional flow cytometry method for assessing the ability of a drug to inhibit Pgp-mediated drug efflux from CR1R12 multidrug-resistant cells has been evaluated. Quantitation of the relative fluorescence was used to compare potency of individual inhibitors. Known Pgp inhibitors, such as cyclosporin A, nicardipine, verapamil, quinidine, terfenadine, tamoxifen, and vinblastine were demonstrated to inhibit the Pgp-mediated efflux of daunorubicin. Cyclosporin A and terfenadine were the most potent inhibitors among the compounds tested. Tetraphenylphosphonium and alpha-tocopherol had little inhibitory effect. Progesterone produced significant inhibition at relatively high concentrations. This study demonstrated that this in vitro flow cytometry method is a simple, sensitive, and quantitative tool to assess the capacity of a drug to inhibit Pgp transporters, and is useful for screening or identifying inhibitors of Pgp as well as evaluation of potential for drug interactions.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Daunorubicin hydrochloride, meets USP testing specifications