Přejít k obsahu
Merck
  • MicroRNA-150 Modulates Adipogenic Differentiation of Adipose-Derived Stem Cells by Targeting Notch3.
MicroRNA-150 Modulates Adipogenic Differentiation of Adipose-Derived Stem Cells by Targeting Notch3.
Stem cells international (2019-11-30)
Xiang Li, Yu Zhao, Xiuquan Li, Qiao Wang, Qiang Ao, Xiaohong Wang, Xiaohong Tian, Hao Tong, Deyu Kong, Shijie Chang, Shuling Bai, Jun Fan
ANOTACE
MicroRNAs (miRNAs) influence stem cell functions, including mobilization, proliferation, and differentiation. miR-150 is abundantly expressed in monocytes. Knockdown of miR-150 promotes bone marrow stem cell migration. The role of miR-150 in adipose-derived stem cells (ADSCs) is unclear. In this study, the effects of miR-150 on adipogenic differentiation and proliferation of ADSCs were investigated. ADSCs were isolated from the inguinal adipose tissue of wild-type (WT) and miR-150 knockout (KO) mice and were induced for adipogenic differentiation. The miR-150 level was detected by real-time PCR. ADSCs were transfected by miR-150 or small-interfering RNA (siRNA) of Notch3. MTT assay and colony formation assay were performed in miR-150 knockdown and control ADSCs. Real-time PCR showed that miR-150 was expressed in ADSCs. miR-150 knockdown significantly decreased the capacity of adipogenic differentiation of ADSCs, as compared with their counterparts from WT mice. It is intriguing that the overexpression of miR-150 significantly increased C/EBPα and PPAR-γ expression and lipid formation in ADSCs with adipogenic induction. Overexpression of miR-150 significantly decreased Notch3 expression in ADSCs compared with the control groups. Furthermore, Notch3 inhibition promoted the adipogenic differentiation in ADSCs. miR-150 also suppressed proliferation potential and the expression of Nanog in ADSCs. In summary, this study demonstrates, for the first time, that miR-150 promotes adipogenic differentiation and inhibits proliferation of ADSCs. miR-150 regulates adipogenic differentiation of ADSCs, likely mediated by the downregulation of Notch3.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Alkaline Phosphatase Diethanolamine Activity Kit, sufficient for 100 tests