Skip To
54987
Holder and Coupler for Legacy HPLC Guard Columns
pkg of 1 ea, for use with non-Supelco Guard Cartridges (not TSKgel Guard Cartridges)
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 ea
Available to ship TODAYfromMILWAUKEE
$273.00
About This Item
UNSPSC Code:
41115700
NACRES:
NB.21
$273.00
Available to ship TODAYDetails
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Product Name
Holder and Coupler for Legacy Guard Columns, for use with non-Supelco guard cartridges (not TSKgel guard cartridges)
compatibility
for use with non-Supelco guard cartridges (not TSKgel guard cartridges)
General description
Finger-tight design couples HPLC columns that use 1/16" 10-32 Valco-compatible fittings with seating distance of 0.074". Also used as a replacement for PEEK coupler supplied in stand-alone guard cartridge holder, Cat. No. 54987.
Other Notes
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
549861.514871.52037
compatibility
for use with non-Supelco guard cartridges (not TSKgel guard cartridges)
compatibility
Also a replacement connector for stand-alone guard holder (54987), for use with HPLC columns with 1/16", 10-32 thread end-fittings (Used with all Supelco and Astec columns, plus other brands that have the same thread dimensions.)
compatibility
-
compatibility
-
Choose from one of the most recent versions:
Certificates of Analysis (COA)