Skip To
22013
SWAGELOK® Back Ferrule
SWAGELOK® 204-1, brass, 1/8 in. SWAGELOK
Synonym(s):
Swagelok® 204-1,BRASS BK FERR 1/8" PK20
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
20 ea
Available to ship TODAYfromMILWAUKEE
$35.40
About This Item
UNSPSC Code:
41115717
NACRES:
SB.54
$35.40
Available to ship TODAYDetails
Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
brass
packaging
pkg of 20 ea
manufacturer/tradename
SWAGELOK® 204-1
fitting
1/8 in. SWAGELOK
General description
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. Glass chromatographic column require brass back ferrule, PTFE front ferrule and brass nuts for adequate sealing.
Legal Information
SWAGELOK is a registered trademark of Swagelok Company
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
220392202822049-U
fitting
1/8 in. SWAGELOK
fitting
1/8 in. SWAGELOK
fitting
1/4 in. SWAGELOK
fitting
1/16 in. SWAGELOK
packaging
pkg of 20 ea
packaging
pkg of 5 ea
packaging
pkg of 5 ea
packaging
pkg of 2 ea
material
brass
material
stainless steel
material
stainless steel
material
stainless steel
manufacturer/tradename
SWAGELOK® 204-1
manufacturer/tradename
SWAGELOK® 204-1
manufacturer/tradename
SWAGELOK® 404-1
manufacturer/tradename
SWAGELOK® 104-1
Still not finding the right product?
Explore all of our products under Swagelok® Back Ferrule
Choose from one of the most recent versions:
Certificates of Analysis (COA)