Skip To
Z360422
Desiccator plate
diam. 230 mm, 24 holes
Synonym(s):
perforated plate, sample support plate, Nalgene 5312-0230, desiccator plate, plate
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
$223.00
6 ea
Available to ship TODAYfromMILWAUKEE
$991.00
About This Item
NACRES:
NB.43
UNSPSC Code:
41121800
$223.00
Check Cart for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
manufacturer/tradename
Nalgene® 5312-0230
O.D.
230 mm
diam.
230 mm
General description
This desiccator plate has a greater resistance to thermal shock than porcelain plates. It has a fire-polished surface, and is a light green ceramic bonded to a metal core for inertness and corrosion-resistance. Numbered quadrants are marked for easy location of crucibles, weighing bottles, and other containers. Recommended use with Nalgene desiccator such as D2547, Z354074 and D2797.
The center hole is 7/8 in. diam. The plates have 24 holes.
Features and Benefits
  • 230mm diameter fits the Nalgene Classic Dessicator (5311-0250 / D2797-1EA) as well as the 250mm Nalgene Blue Desiccators (5309-0250 / D2547-1EA and 5310-0250 / Z354074-1EA).
  • Numbered quadrants for easy location of crucibles and other containers.
  • Six holes in each quadrant and a 3mm center hole to enhance airflow and drying efficiency.
  • Autoclavable.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
CLS353503WHA77009901WHA77009904
O.D.
230 mm
O.D.
-
O.D.
-
O.D.
-
manufacturer/tradename
Nalgene® 5312-0230
manufacturer/tradename
Corning 353503
manufacturer/tradename
Whatman 7700-9901, Whatman Article No. 28420845 (US reference)
manufacturer/tradename
Whatman 7700-9904, Whatman Article No. 28420848 (US reference)
diam.
230 mm
diam.
-
diam.
-
diam.
-
Choose from one of the most recent versions:
Certificates of Analysis (COA)