Skip To
T1418
Nalgene® centrifuge tubes, Oak Ridge Style 3119
nominal capacity 50 mL
Synonym(s):
High-Speed ​​Centrifuge Tubes, Nalgene 3119-0050, sample collection tube, sample receptacle, sample tube
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
10 ea
Check Cart for Availability
$211.00
About This Item
NACRES:
NB.11
UNSPSC Code:
41121703
Material:
polypropylene copolymer , polypropylene screw closure, round bottom tube, screw top cap (Linerless)
Sterility:
non-sterile
Packaging:
pack of 10 ea
Feature:
RCF 100,605 × g
$211.00
Check Cart for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene copolymer , polypropylene screw closure, round bottom tube, screw top cap (Linerless)
sterility
non-sterile
feature
RCF 100,605 × g
packaging
pack of 10 ea
manufacturer/tradename
Nalgene® 3119-0050
nominal capacity
50 mL
General description
“Nalgene” Oak Ridge high-speed polypropylene copolymer (PPCO) centrifuge tubes offer broader chemical compatibility with lab reagents compared to polycarbonate centrifuge tubes.
Features and Benefits
  • Oak Ridge style centrifuge tubes are strong with excellent mechanical strength.
  • Chemically resistant and can withstand repeated autoclave cycles.
  • Clear tubes allow for optimal sample visibility.
  • Polypropylene screw closure provides a leak-proof seal to prevent sample loss.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
T1043T1293T1168
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
-
material
polypropylene copolymer , round bottom tube, polypropylene screw closure, screw top cap (Linerless)
material
polypropylene copolymer , polypropylene screw closure, round bottom tube, screw top cap (Linerless)
material
polypropylene copolymer , polypropylene screw closure, round bottom tube, screw top cap (Linerless)
material
polypropylene copolymer , polypropylene screw closure, round bottom tube, screw top cap (Linerless)
manufacturer/tradename
Nalgene® 3119-0050
manufacturer/tradename
Nalgene® 3119-0010
manufacturer/tradename
Nalgene® 3119-0030
manufacturer/tradename
Nalgene® 3119-0028
packaging
pack of 10 ea
packaging
pack of 10 ea
packaging
pack of 10 ea
packaging
pack of 10 ea
feature
RCF 100,605 × g
feature
RCF 50,000 × g
feature
RCF 50,000 × g
feature
RCF 50,000 × g
nominal capacity
50 mL
nominal capacity
10 mL
nominal capacity
30 mL
nominal capacity
28 mL
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)