Skip To
L8144
Sigma label tape
blue
Synonym(s):
Sigma tape
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
1 ea
Available to ship TODAYfromMILWAUKEE
$31.40
5 ea
Available to ship TODAYfromMILWAUKEE
$157.00
About This Item
NACRES:
NB.35
UNSPSC Code:
55121611
Manufacturer/tradename:
Sigma-Aldrich
Material:
blue
Packaging:
pkg of 1 roll
$31.40
Available to ship TODAYDetails
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
blue
packaging
pkg of 1 roll
manufacturer/tradename
Sigma-Aldrich
W
3/4 in. (1.91 cm)
core size
3 in. (7.62 cm)
roll size
2160 in. (55 m)
General description
All purpose, self-adhesive tape can be written on with pen, pencil or permanent marker. Temperatures from -23°C to +121°C will not affect tape.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
L8269L8394L8519
material
blue
material
green
material
white
material
red
packaging
pkg of 1 roll
packaging
pkg of 1 roll
packaging
pkg of 1 roll
packaging
pkg of 1 roll
core size
3 in. (7.62 cm)
core size
3 in. (7.62 cm)
core size
3 in. (7.62 cm)
core size
3 in. (7.62 cm)
W
3/4 in. (1.91 cm)
W
3/4 in. (1.91 cm)
W
3/4 in. (1.91 cm)
W
3/4 in. (1.91 cm)
roll size
2160 in. (55 m)
roll size
2160 in. (55 m)
roll size
2160 in. (55 m)
roll size
2160 in. (55 m)
Still not finding the right product?
Explore all of our products under Sigma label tape
Choose from one of the most recent versions:
Certificates of Analysis (COA)