Skip To
CLS4515
Corning® spheroid microplates
96 well, round black/clear bottom, ULA (Ultra-Low Attachment surface), lid, sterile
Synonym(s):
96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate, EIA/RIA plate, immunoassay plate
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
5 ea
Check Cart for Availability
$209.00
About This Item
NACRES:
NB.13
UNSPSC Code:
41121800
Material:
clear bottom, polystyrene , round black/clear bottom
Size:
96 wells
$209.00
Check Cart for Availability
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
clear bottom, polystyrene , round black/clear bottom
sterility
sterile
feature
lid, skirt, Specific plates for 3D cell culture, ULA (Ultra-Low Attachment surface)
packaging
pkg of 5 ea
manufacturer/tradename
Corning 4515
size
96 wells
well volume
300 μL
wells
96 wells
working volume
0.075-0.30 μL
suitability
suitable for (cell cuture)
binding type
(Ultra-Low Attachment)
General description
Key Features:
  • Optically clear round bottom with black opaque microplate body
  • Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
  • Novel proprietary well geometry aids formation of spheroids in center of well
  • Unique design that shields each well to minimize well-to-well cross talk
Key Benefits:
  • Uniform, single spheroid formation across all wells
  • Centering of the spheroid in each well enables automated visualization
  • Culture and assay spheroids in the same microplate, without the need for transfer to a new plate
  • Ease of media exchange or treatment with test compounds/drugs
Legal Information
Corning is a registered trademark of Corning, Inc.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
CLS4516CLS4527CLS3830
suitability
suitable for (cell cuture)
suitability
suitable for (cell cuture)
suitability
suitable for (cell cuture)
suitability
suitable for (cell cuture)
size
96 wells
size
384 wells
size
1536 wells
size
384 wells
material
clear bottom, round black/clear bottom , polystyrene
material
clear bottom, polystyrene , round black/clear bottom
material
polystyrene , round black/clear bottom
material
clear bottom, polystyrene , round black/clear bottom
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
feature
lid, Specific plates for 3D cell culture, skirt, ULA (Ultra-Low Attachment surface)
feature
lid, skirt, Specific plates for 3D cell culture, ULA (Ultra-Low Attachment surface)
feature
ULA (Ultra-Low Attachment surface), Lid Included, Recommended Medium Well Volume, Recommended Working Volume, Specific plates for 3D cell culture
feature
lid, skirt, Specific plates for 3D cell culture, ULA (Ultra-Low Attachment surface)
well volume
300 μL
well volume
90 μL
well volume
(14 mL)
well volume
90 μL
Still not finding the right product?
Explore all of our products under Corning® spheroid microplates
Choose from one of the most recent versions:
Certificates of Analysis (COA)