Skip To
CLS3295
Corning® CellBIND® surface culture dishes
D × H 60 mm × 15 mm
Synonym(s):
cell culture flasks, tissue culture flasks
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
126 ea
Available to ship TODAYfromMILWAUKEE
$348.00
About This Item
NACRES:
NB.13
UNSPSC Code:
41121812
Material:
polystyrene , round
Sterility:
sterile; γ-irradiated
Manufacturer/tradename:
Corning 3295
Packaging:
sleeve of 7
$348.00
Available to ship TODAYDetails
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polystyrene , round
sterility
sterile; γ-irradiated
packaging
sleeve of 7
manufacturer/tradename
Corning 3295
D × H
60 mm × 15 mm
surface area
21 cm2
General description
  • Manufactured from optically-clear virgin polystyrene
  • Corning® CellBIND® surface treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Supplied with vents to provide consistent gas exchange
Legal Information
CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
CLS3296CLS430597CLS3294
manufacturer/tradename
Corning 3295
manufacturer/tradename
Corning 3296
manufacturer/tradename
Corning 430597
manufacturer/tradename
Corning 3294
sterility
sterile; γ-irradiated
sterility
sterile; γ-irradiated
sterility
sterile
sterility
sterile; γ-irradiated
material
polystyrene , round
material
polystyrene , round
material
polystyrene , round
material
polystyrene , round
surface area
21 cm2
surface area
55 cm2
surface area
148 cm2
surface area
8 cm2
D × H
60 mm × 15 mm
D × H
-
D × H
150 mm × 25 mm
D × H
35 mm × 10 mm
packaging
sleeve of 7
packaging
sleeve of 5
packaging
sleeve of 5
packaging
sleeve of 10
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)