Skip To
CLS3013
Corning® single use spatulas
microspatula, rounded end/scoop
Synonym(s):
Spatula for Lab Use
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
50 ea
Available to ship TODAYfromMILWAUKEE
$170.00
About This Item
NACRES:
NB.14
UNSPSC Code:
41122403
$170.00
Available to ship TODAYDetails
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
sterility
sterile
manufacturer/tradename
Corning 3013
L
150 mm
General description
Scoop dimensions:
Maximum scoop width - 0.13 inches (3.302 mm)
scoop can hold < 0.5 mL
Corning single use spatulas have been designed to save time and provide contamination-free samples. Eliminates need to recycle and sterilize reusable spatulas.
Each spatula is individually packaged, RNase/DNase free, non-pyrogenic and antistatic.
Legal Information
Corning is a registered trademark of Corning, Inc.
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
CLS3012CLS3006CLS3007
L
150 mm
L
150 mm
L
250 mm
L
250 mm
manufacturer/tradename
Corning 3013
manufacturer/tradename
Corning 3012
manufacturer/tradename
Corning 3006
manufacturer/tradename
Corning 3007
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
Still not finding the right product?
Explore all of our products under Corning® single use spatulas
Choose from one of the most recent versions:
Certificates of Analysis (COA)