Skip To
B8032
Nalgene® narrow mouth PPCO bottle
polypropylene copolymer, capacity 30 mL, translucent, with cap
Synonym(s):
nalgene bottle, nalgene bottles, Nalgene 2006-0001, laboratory container, receptical, vessel
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
12 ea
Available to ship TODAYfromMILWAUKEE
$36.90
About This Item
NACRES:
NB.17
UNSPSC Code:
41121800
$36.90
Available to ship TODAYDetails
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene cap, translucent , polypropylene copolymer , polypropylene copolymer bottle (PPCO)
joint
Joints threaded neck (20)
description
Autoclavable
feature
with cap
packaging
pack of 12 ea
manufacturer/tradename
Nalgene® 2006-0001
bottle diam. × H
34 mm × 61 mm
capacity
30 mL
General description
For lab applications requiring excellent chemical resistance and autoclaving, with and without contents, use Nalgene®arrow-mouth PPCO bottles with Closure. These polypropylene copolymer bottles are translucent, providing good clarity for viewing contents. Made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables.
Features and Benefits
  • Nalgene® narrow-mouth bottles are ideal for long-term storage.
  • Made from polypropylene copolymer (PPCO) material for excellent sample clarity.
  • Highly chemical resistant to most acids, bases, and alcohols.
  • Autoclavable - with and without contents.
  • Laboratory-grade plastic materials for dependably low leachables and extractables.
  • Liner less caps ensures leak-proof seal for sample protection.
  • Light-weight - enables easy lifting and transporting.
  • Avoid UV exposure.
  • Manufactured from non-toxic materials.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
B8157B8407B8282
feature
with cap
feature
with cap
feature
with cap
feature
with cap
material
polypropylene cap, polypropylene copolymer bottle (PPCO), translucent
material
polypropylene copolymer , polypropylene copolymer bottle (PPCO), round translucent bottle
material
polypropylene copolymer , polypropylene copolymer bottle (PPCO), round bottle, translucent
material
polypropylene copolymer , polypropylene copolymer bottle (PPCO), round bottle, translucent
manufacturer/tradename
Nalgene® 2006-0001
manufacturer/tradename
Nalgene® 2006-0002
manufacturer/tradename
Nalgene® 2006-0008
manufacturer/tradename
Nalgene® 2006-0004
capacity
30 mL
capacity
60 mL
capacity
250 mL
capacity
125 mL
joint
Joints threaded neck (20)
joint
Joints threaded neck (20)
joint
Joints threaded neck (24)
joint
Joints threaded neck (24)
packaging
pack of 12 ea
packaging
pack of 12 ea
packaging
pack of 12 ea
packaging
pack of 12 ea
Still not finding the right product?
Explore all of our products under Nalgene® narrow mouth PPCO bottle
Choose from one of the most recent versions:
Certificates of Analysis (COA)