Skip to Content
MilliporeSigma

227039

Cholera Toxin, B Subunit, Vibrio cholerae, Type Inaba 569B - Calbiochem

Subunit of cholera toxin responsible for binding to GM1 ganglioside receptors in membranes.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

solubility

sterile distilled water: soluble

shipped in

ambient

storage temp.

2-8°C

General description

Cholera toxin is an oligomeric protein consisting of a single A subunit (M.W. 28,000) containing two polypeptide chains, A1 and A2 , and a B subunit (M.W. 55,000) containing five polypeptide chains. Cholera toxin is the pathogenic agent responsible for the symptoms of cholera. Cholera toxin is also a potent activator of adenylate cyclase that binds the ADP-ribosylation factor and modulates its activity. The A subunit catalyzes the ADP-ribosylation of GS that leads to the persistent activation of adenylate cyclase. The consequent increase in intracellular cAMP levels stimulates Cl- secretion and leads to severe dehydration. The B subunit binds specifically to GM1 ganglioside receptors, which are expressed ubiquitously in mammalian cells, and assists in the transport of the A subunit through the membrane. When used alone, the B subunit will block the action of toxin on intact cells. Purified B subunit is produced by a modification of the methods published by Lai, C.Y., et al.
Subunit of cholera toxin responsible for binding to GM1 ganglioside receptors in membranes. When used along, will block the action of toxin on intact cells.

Biochem/physiol Actions

Cell permeable: no
Cholera Toxin, B Subunit is tested for its effectiveness in binding antitoxin using the antitoxin combining power assay described by Craig, J.P., et al.
Primary Target
GM1 ganglioside receptors
Reversible: no

Packaging

yes

Physical form

Lyophilized from 200 mM NaCl, 50 mM Tris-HCl, 3 mM NaN₃, 1 mM EDTA, pH 7.5.

Preparation Note

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 6 months at 4°C. DO NOT FREEZE reconstituted solutions.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
McCloskey, M.A. and Zhang, L. 2000. J. Cell. Biol.148, 137.
Mulhein, S.A., et al. 1989. J. Membr. Biol. 109, 21.
Tsai, S.-C., et al. 1988. J. Biol. Chem.263, 1768.
Van Heynigen, S. 1982. Biosci. Rep.2, 135.
Mekalanos, J.J., et al. 1978. Infect. Immun.20, 552.
Gill, D.M. 1976. Biochemistry15, 1242.
Lai, C.Y., et al. 1976. J. Infect. Dis.139, 674.
Rappaport, R.S., et al. 1974. Infect. Immun.9, 294.
Craig, J.P. 1971. In Kadies, S., et al. (Eds.). Microbial Toxins. New York. Academic Press, 2A, 189.

Legal Information

Not available for sale outside of the United States.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Highly Toxic (H)


pictograms

Exclamation mark

signalword

Warning



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library