Skip to Content
MilliporeSigma

04-1660

Rabbit Anti-Human Ig kappa light chain Antibody, clone RM126

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
RM126, monoclonal
Species reactivity:
human, rabbit
Application:
ELISA, IHC
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody form

purified antibody

Quality Segment

clone

RM126, monoclonal

purified by

using Protein A

species reactivity

human, rabbit

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable, immunohistochemistry: suitable

isotype

IgG

General description

Human IgG

Application

Immunohistochemistry Analysis (IHC): 0.5-2 μg/mL from a representative lot detected human Ig kappa light chain in human tonsil tissue.

Physical form

Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

ELISA Analysis: The specificity and reactivity of a representative lot was confirmed by ELISA analyses.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library