Key Documents
Z611263
Hirschmann® microcapillary pipette
tube capacity 100 μL
About This Item
UNSPSC Code:
41121500
eCl@ss:
32070100
NACRES:
NB.01
material
blue
glass
description
Genuine Schott® Duran glass
packaging
pack of 250 ea
manufacturer/tradename
Hirschmann 9600199
availability
not available in USA and Canada
L
125 mm
tube capacity
100 μL
General description
Disposable pipettes with ringmark. The exact ringmark guarantees an accurate volume determination. Each micro capillary is color-coded to avoid mix ups. Useful for spotting TLC plates.
Legal Information
Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.
Schott is a registered trademark of Schott AG