Key Documents
Z413380
Sigma-Aldrich
Aldrich® Hempel column
Unjacketed, effective L 300 mm, O.D. × I.D. 22 mm × 19 mm
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
joint
joint ST/NS24/40
O.D. × I.D.
22 mm × 19 mm
effective L
300 mm