Key Documents
Z283274
Sigma-Aldrich
Aldrich® heavy-duty spatula
L 12 in.
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
material
stainless steel
L
12 in.
General description
Double flat ends; one rounded, one square. Each end is 3 L × 3/4 in. W.
Application
Aldrich® heavy-duty spatula has been used:
  • to scrape the cells during the collection of E. coli cell culture samples
  • to add the sand to the stainless steel tube
  • to pry and mount the blocks of organoids