GC Analysis of Volatiles in Green Arabica Coffee on SUPELCOWAX 10 after SPME using 50/30 μm DVB/Carboxen/PDMS Fiber
CONDITIONS
sample/matrix
Arabica coffee beans, green, 2 grams with pre-equilibration of 10 min at 60 °C, the entire process under agitation at 500 rpm
SPME fiber
divinylbenzene/carboxen on polydimethylsiloxane, 50/30 μm (57328-U)
extraction
headspace, 40 min at 60 °C
desorption process
5 min
column
SUPELCOWAX 10, 30 m × 0.25 mm I.D., 0.25 μm (24079)
oven
40 °C (5 min), 4 °C/min to 230 °C, 50 °C/min to 280°C (2 min)
inj. temp.
260 °C
detector
MS (Shimadzu GCMS-QP2010) ionization energy 1.5 kV; mass range: 40-400 amu; scan mode, scan interval : 0.5 sec
MSD interface
230 °C, source temp. 200 °C
carrier gas
helium, 34.0 cm/s and 40.3 KPa
injection
splitless 5 min, then split ratio 1:20
liner
0.75 mm i.d.
Description
Legal Information
null