GC Analysis of 1-Aminoindan Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ G-DP
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Astec CHIRALDEX G-DP, 30 m x 0.25 mm I.D., 0.12 μm (78033AST)
oven
160 °C
inj. temp.
250 °C
carrier gas
helium, 30 psi
sample
peaks 1 & 2: 1-aminoindan enantiomers (N-TFA derivatives)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC