Merck
  • Genetic derangements in the tumor suppressor gene PTEN in endometrial precancers as prognostic markers for cancer development: a population-based study from northern Norway with long-term follow-up.
Genetic derangements in the tumor suppressor gene PTEN in endometrial precancers as prognostic markers for cancer development: a population-based study from northern Norway with long-term follow-up.
Gynecologic oncology (2004-09-24)
A Orbo, T Kaino, M Arnes, M Kopp, K Eklo
ABSTRACT
The purpose of the current study was to characterize the role of PTEN in malignant transformation and to evaluate the significance of mutated PTEN exons as prognostic markers in the carcinogenesis of endometrial hyperplasia. A comparison of PTEN mutations as prognostic markers with former investigated prognosticators was also intended. Histological material from 68 patients with endometrial hyperplasia and 10-20 years of follow-up of whom 18 later developed cancer was examined. PCR amplification and DNA sequencing were performed, screening the most frequently mutated exons 5a-8b of the PTEN gene. Mutations were demonstrated in 13.2% of the patients. Of the patients with cancer development, five showed to have PTEN mutations corresponding to 28%. Of the patients remaining without carcinoma, only 8% had PTEN mutations (P = 0.04). In total, there were three missense, three nonsense, and four frameshift mutations, and twice as many mutations leading to a truncated protein (six) than mutations altering one amino acid in the entire protein (three). Mutations were distributed in the following manner: three in exon 5a, two in exon 5b, two in exon 6, two in exon 7, and one in exon 8b. Only mutations in exons 6, 7, and 8a were connected with cancer development or coexisting cancer and six out of seven mutations within these exons were frameshift or nonsense mutations. Our results showed that mutations in the PTEN gene were statistically more frequent in cases with cancer development or coexisting cancer. Although the specificity was acceptable, the sensitivity of PTEN mutations was too low to make it suitable as a tumor marker (sensitivity of 27% and specificity of 91%) in clinical practice.
Product Description
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate