Merck
ADAMTS-1 Is Found in the Nuclei of Normal and Tumoral Breast Cells.
PloS one (2016-10-21)
Suély V Silva, Maíra A Lima, Nathalie Cella, Ruy G Jaeger, Vanessa M Freitas
ABSTRACT
Proteins secreted in the extracellular matrix microenvironment (ECM) by tumor cells are involved in cell adhesion, motility, intercellular communication and invasion. The tumor microenvironment is expansively modified and remodeled by proteases, resulting in important changes in both cell-cell and cell-ECM interactions and in the generation of new signals from the cell surface. Metalloproteinases belonging to the ADAMTS (a disintegrin and metalloprotease with thrombospondin motifs) family have been implicated in tissue remodeling events observed in cancer development, growth and progression. Here we investigated the subcellular localization of ADAMTS-1 in normal-like (MCF10-A) and tumoral (MCF7 and MDA-MB-231) human breast cells. ADAMTS-1 is a secreted protease found in the extracellular matrix. However, in this study we show for the first time that ADAMTS-1 is also present in the nuclei and nucleoli of the three mammary cell lines studied here. Our findings indicate that ADAMTS-1 has proteolytic functions in the nucleus through its interaction with aggrecan substrate.
Sigma-Aldrich
Anti-Aggrecan (Cleaved-Asp369), N-Terminal antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Histone H1 Antibody, clone AE-4, clone AE-4, Upstate®, from mouse
Sigma-Aldrich
Anti-ADAMTS-1 Antibody, clone 5D4E11B6, serum, clone 5D4E11B6, Chemicon®
Sigma-Aldrich
Anti-B23 antibody, Mouse monoclonal, clone FC82291, purified from hybridoma cell culture