Merck
  • Semiautomated determination of neonicotinoids and characteristic metabolite in urine samples using TurboFlow™ coupled to ultra high performance liquid chromatography coupled to Orbitrap analyzer.
Semiautomated determination of neonicotinoids and characteristic metabolite in urine samples using TurboFlow™ coupled to ultra high performance liquid chromatography coupled to Orbitrap analyzer.
Journal of pharmaceutical and biomedical analysis (2017-09-18)
Marina López-García, Roberto Romero-González, Marina Lacasaña, Antonia Garrido Frenich
ABSTRACT
A semiautomated method based on ultra-high performance liquid chromatography (UHPLC) coupled to Orbitrap high resolution mass spectrometry has been developed for the determination of neonicotinoids (imidacloprid, acetamiprid, clothianidin, dinotefuran, nitenpyram, thiacloprid and thiamethoxam) and the metabolite acetamiprid-n-desmethyl in urine samples. Two automated methods were tested (solid-phase extraction "SPE" and turbulent flow chromatography "TurboFlow™"), obtaining the best results when TurboFlow™ was applied. The total analysis time for the developed method was 14min. The optimized method was validated, obtaining suitable results for all validation parameters. Recoveries ranged from 78% to 116% meanwhile repeatability and reproducibility were evaluated obtaining values lower than 10% and 20% respectively (except for dinotefuran and nitenpyram at 0.2μgL-1). The limit of quantification (LOQ) for all compounds was established at 0.2μgL-1. The proposed analytical methodology was applied to analyze the target compounds in thirty six urine samples from pregnant women living in agricultural areas of Almería (Spain). Imidacloprid, acetamiprid and acetamiprid-n-desmethyl were detected in some of the samples at concentrations ranging from 0.23 to 1.57μgL-1. Furthermore, dinotefuran was identified in two samples at trace levels.
Product Description
Supelco
Thiacloprid, PESTANAL®, analytical standard
Sigma-Aldrich
Phe-Ala
Supelco
Acetamiprid-N-desmethyl, PESTANAL®, analytical standard
Supelco
Thiamethoxam, PESTANAL®, analytical standard
Supelco
N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine, PESTANAL®, analytical standard
Supelco
Clothianidin, PESTANAL®, analytical standard
Supelco
Nitenpyram, PESTANAL®, analytical standard