Merck
CO
Theranostics (2017-12-01)
Yuanyuan Li, Shunjie Liu, Xun Zhao, Ying Wang, Jianhua Liu, Xianhong Wang, Lehui Lu
ABSTRACT
Biodegradable polymeric nanomaterials can be directly broken down by intracellular processes, offering a desirable way to solve toxicity issues for cancer diagnosis and treatment. Among them, aliphatic polycarbonates are approved for application in biological fields by the United States Food and Drug Administration (FDA), however, high hydrophobicity, deficient functionality and improper degradation offer significant room for improvement in these materials. To achieve progress in this direction, herein, we demonstrate that CO The resulting polymeric micelles (APC-DTPA/Gd) exhibit excellent magnetic resonance imaging performance, simultaneously enabling real-time visualization of bioaccumulation and decomposition of polymeric micelles Our findings may help the development of CO
Sigma-Aldrich
2,2-Dimethoxy-2-phenylacetophenone, 99%
Sigma-Aldrich
Diethylenetriaminepentaacetic dianhydride, 98%